Skip to Content
Merck
All Photos(1)

Key Documents

56581

Supelco

Sodium perchlorate monohydrate

suitable for HPLC, LiChropur, 99.0-101.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO4 · H2O
CAS Number:
Molecular Weight:
140.46
MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21
Pricing and availability is not currently available.

Assay

99.0-101.0% (T)

form

solid

manufacturer/tradename

LiChropur

technique(s)

HPLC: suitable

pH

4.5-8.0

solubility

H2O: 5%, clear, colorless

λ

10 % in water

UV absorption

λ: 210 nm Amax: ≤0.1
λ: 220 nm Amax: ≤0.05
λ: 230 nm Amax: ≤0.03
λ: 240 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

SMILES string

O.[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1

InChI key

IXGNPUSUVRTQGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium perchlorate monohydrate along with acetonitrile may be used as a mobile phase in:
  • Determination of methenamine, methenamine mandelate and methenamine hippurate in pharmaceutical preparations using ion-exchange high-performance liquid chromatography (HPLC).[1]
  • Development and validation of RP-HPLC method for the dissolution and assay of etoricoxib in pharmaceutical dosage forms.[2]
  • Determination of nitrate and nitrite by HPLC in human plasma.[3]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of nitrate and nitrite by high-performance liquid chromatography in human plasma
Jedlickova V, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 780(1), 193-197 (2002)
Development and validation of RP-HPLC method for the dissolution and assay of etoricoxib in pharmaceutical dosage forms
Singh B, et al.
International Journal of Pharmaceutical Quality Assurance, 6(01), 1-7 (2015)
Chiravi Pavitrapok et al.
Journal of pharmaceutical and biomedical analysis, 40(5), 1243-1248 (2005-11-22)
An ion-exchange column high-performance liquid chromatography (HPLC) method has been developed for the determination of methenamine in methenamine and methenamine hippurate pharmaceutical preparations. The HPLC method uses a Zorbax SCX-300 column with acetonitrile-0.1M sodium perchlorate monohydrate (pH 5.8) (70:30, v/v)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service