Skip to Content
Merck
All Photos(2)

Key Documents

28660

Supelco

Procyanidin A2

analytical standard

Synonym(s):

(+)-Epicatechin-(4β-8,2β-O-7)-epicatechin, Proanthocyanidin A2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24O12
CAS Number:
Molecular Weight:
576.50
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1[C@@H](C2=CC(O)=C(O)C=C2)OC3=C4[C@@]5([H])C6=C(O)C=C(O)C=C6O[C@](OC4=CC(O)=C3C1)(C7=CC(O)=C(O)C=C7)[C@@H]5O

InChI

1S/C30H24O12/c31-13-7-20(37)24-22(8-13)41-30(12-2-4-16(33)19(36)6-12)29(39)26(24)25-23(42-30)10-17(34)14-9-21(38)27(40-28(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,21,26-27,29,31-39H,9H2/t21-,26-,27-,29-,30+/m1/s1

InChI key

NSEWTSAADLNHNH-LSBOWGMISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Procyanidins are oligomers and polymers of flavan-3-ols consisting of (−)-epicatechin subunits found in fruits and plants with various biological benefits. Procyanidin A2 is a dimer of procyanidin.

Application

Procyanidin A2 may be used as a reference standard for the quantification of the analyte in grapes and wines of Vitis vinifera L. of Cabernet Sauvignon and Merlot noir by reverse-phase high-performance liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: crataegus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of procyanidin A2 in grape and wine of Vitis vinifera L. cv: Merlot noir and Cabernet Sauvignon.
de Gaulejac NV, et al.
International Journal of Grape and Wine Science, 35(1), 51-56 (2001)
Variations in the heterogeneity of the decay of the fluorescence in six procyanidin dimers.
Cho D, et al.
Journal of the American Chemical Society, 112(11), 4273-4277 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service