Skip to Content
Merck
All Photos(1)

Key Documents

1.05866

Supelco

Magnesium oxide

for analysis (max.0.001% SO₄) EMSURE® ACS

Synonym(s):

Magnesium oxide, Magnesia usta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO
CAS Number:
Molecular Weight:
40.30
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-171-9
NACRES:
NA.21
Assay:
≥97% (complexometric)
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥97% (complexometric)

form

solid

impurities

≤0.02% Substances insoluble in hydrochloric acid
≤0.02% Substances precipitated by ammonia
≤0.4% Substances soluble in water

loss

≤2% loss on ignition, 1000 °C

pH

10.3 (20 °C in H2O, saturated solution)

bp

3600 °C/1000 hPa

mp

2800 °C

density

3.58 g/cm3 at 25 °C

bulk density

100 kg/m3

anion traces

carbonate (CO32-): ≤1.5%
chloride (Cl-): ≤0.01%
nitrate (NO3-): ≤0.005%
sulfate (SO42-): ≤0.001%
sulfate, sulfite (as SO42-): ≤0.02%

cation traces

As: ≤0.0001%
Ba + Sr (as Ba): ≤0.005%
Ca: ≤0.05%
Cu: ≤0.001%
Fe: ≤0.005%
K: ≤0.005%
Mn: ≤0.0005%
N: ≤0.002%
Na: ≤0.25%
Pb: ≤0.001%
Zn: ≤0.0005%
heavy metals (as Pb): ≤0.003%

storage temp.

no temp limit

SMILES string

[Mg]=O

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Related Categories

Analysis Note

Identity: passes test
Assay (complexometric): ≥ 97 %
Appearance of solution: passes test
Substances soluble in water: ≤ 0.4 %
Substances insoluble in hydrochloric acid: ≤ 0.02 %
Substances precipitated by ammonia: ≤ 0.02 %
Carbonate (as CO₂): ≤ 1.5 %
Chloride (Cl): ≤ 0.01 %
Nitrate (NO₃): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.001 %
Sulfate and sulphite (as SO₄): ≤ 0.02 %
Total nitrogen (N): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.003 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.05 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.005 %
K (Potassium): ≤ 0.005 %
Mn (Manganese): ≤ 0.0005 %
Na (Sodium): ≤ 0.25 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.0005 %
Barium and Strontium (as Ba): ≤ 0.005 %
Loss on ignition (1000 °C): ≤ 2.0 %
Conforms to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service