Skip to Content
Merck
All Photos(2)

Key Documents

900117P

Avanti

PGE2

Avanti Research - A Croda Brand

Synonym(s):

Prostaglandin E2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
Molecular Weight:
352.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (900117P-10MG)
pkg of 1 × 5 mg (900117P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1CC([C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/[C@@H](O)CCCCC)=O

Packaging

2 mL Amber Glass Sealed Ampule (900117P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meijing Liu et al.
Antioxidants & redox signaling, 33(2), 87-116 (2020-02-11)
Aims: Acetaminophen (APAP) overdose leads to acute liver injury by inducing hepatic mitochondrial oxidative stress and inflammation. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service