Skip to Content
Merck
All Photos(1)

Key Documents

860816P

Avanti

16:0(2S-OH) Ceramide

Avanti Research - A Croda Brand 860816P, powder

Synonym(s):

N-(2′-(S)-hydroxypalmitoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H67NO4
CAS Number:
Molecular Weight:
553.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860816P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860816P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@@H](O)CCCCCCCCCCCCCC)=O)CO

InChI

1S/C34H67NO4/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-32(37)31(30-36)35-34(39)33(38)29-27-25-23-21-18-16-14-12-10-8-6-4-2/h26,28,31-33,36-38H,3-25,27,29-30H2,1-2H3,(H,35,39)/b28-26+/t31-,32+,33-/m0/s1

InChI key

YWZGQPJRQXHVQX-UMKNRRHWSA-N

General description

Ceramides with 2-hydroxy fatty acids (hFA) group are present in the epidermis. 16:0(2S-OH) Ceramide is a unique ceramide containing 16C long chain base fatty acid (palmitic acid)-with 2′-hydroxyl group in S configuration.

Biochem/physiol Actions

Hydroxy fatty acid (hFA)-sphingolipids are implicated in cell signaling. They also facilitate cell differentiation and apoptosis. hFA-ceramides in epidermis help in permeability barrier function. hFA-ceramides are synthesized by fatty acid 2-hydroxylase (FA2H). Mutations of FA2H leads to the development of neurological disorders including leukodystrophy and spastic paraparesis in humans. hFA-ceramides in the cell membrane are essential for PM02734 drug to exhibit its anti-tumor activity.

Packaging

5 mL Amber Glass Screw Cap Vial (860816P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Venkatesh Kota et al.
Advances in biological regulation, 54, 223-230 (2013-10-22)
Ceramide is a precursor of complex sphingolipids and also plays important roles in cell signaling. With the advances in lipid analytical technologies, the structural diversity of ceramide species have become evident, and the complexity of cellular metabolism and function associated
2?-Hydroxy ceramide in membrane homeostasis and cell signaling
Kota V and Hama H
Advances in Biological Regulation, 54, 223-230 (2014)
Levels of SCS7/FA2H-mediated fatty acid 2-hydroxylation determine the sensitivity of cells to antitumor PM02734
Herrero AB, et al.
Cancer research, 68(23), 9779-9787 (2008)
Fatty acid 2-hydroxylase regulates cAMP-induced cell cycle exit in D6P2T schwannoma cells
Alderson NL, et al.
Journal of Lipid Research, 50(6), 1203-1208 (2009)
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology
Hama H, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 405-414 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service