Skip to Content
Merck
All Photos(1)

Key Documents

860692P

Avanti

C16:0 Ceramide (d18:2(4E,8Z)/16:0)

Avanti Research - A Croda Brand 860692P, powder

Synonym(s):

N-palmitoyl-D-erythro-sphingosine (d18:2(4E,8Z) base)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H65NO3
CAS Number:
Molecular Weight:
535.88
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860692P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860692P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](NC(CCCCCCCCCCCCCCC)=O)([H])[C@]([H])(O)/C=C/CC/C=C\CCCCCCCCC

General description

C16:0 Ceramide (d18:2(4E,8Z)/16:0) or N-palmitoyl-D-erythro-sphingosine (d18:2(4E,8Z) base) is a long chain ceramide derived from sphingosine containing two double bonds with palmitic acid. C16:0 Ceramide synthesis is catalyzed by the enzyme ceramide synthase 6 (CerS6).

Biochem/physiol Actions

C16:0-ceramide facilitates obesity-related insulin resistance. It exhibits pro-apoptotic function. Elevated levels of C16:0-ceramide has been observed in adipose tissue of obese patients.

Packaging

5 mL Amber Glass Screw Cap Vial (860692P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah M Turpin et al.
Cell metabolism, 20(4), 678-686 (2014-10-09)
Ceramides increase during obesity and promote insulin resistance. Ceramides vary in acyl-chain lengths from C14:0 to C30:0 and are synthesized by six ceramide synthase enzymes (CerS1-6). It remains unresolved whether obesity-associated alterations of specific CerSs and their defined acyl-chain length
Timothy Hla et al.
Cell metabolism, 20(5), 703-705 (2014-12-03)
A substantive literature has accumulated implicating sphingolipids, in particular ceramides, as mediators of insulin resistance in metabolic syndrome. Thanks to recent technical advances in mouse genetics and lipidomics, two independent laboratories identify the same sphingolipid, C16:0-ceramide, as principal mediator of obesity-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service