Skip to Content
Merck
All Photos(1)

Key Documents

Z308595

Quickfit® Drechsel bottles

capacity 500 mL

Synonym(s):

MF29/3/500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

Joint

joint 24/29

manufacturer/tradename

Quickfit MF 29/3/500

availability

not available in USA and Canada

H

245 mm

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

General description

Select bottle and corresponding head. Note different bottles sizes are different heights.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service