Skip to Content
Merck
All Photos(1)

Key Documents

W368008

Sigma-Aldrich

2-Ethyl-4-methylthiazole

≥98%, FG

Synonym(s):

2-ethyl-4-methyl-1,3-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NS
CAS Number:
Molecular Weight:
127.21
FEMA Number:
3680
EC Number:
Council of Europe no.:
11612
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.033
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets requirements for JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.505 (lit.)

bp

161-162 °C (lit.)

density

1.026 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; nutty

SMILES string

CCc1nc(C)cs1

InChI

1S/C6H9NS/c1-3-6-7-5(2)4-8-6/h4H,3H2,1-2H3

InChI key

VGRVKVGGUPOCMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Ethyl-4-methylthiazoleis a heterocyclic compound belonging to the thiazole family of molecules and is commonly used as a flavoring and fragrance agent . Two-ethyl-4-methylthiazole can be found in coffee. This flavoring is used to flavor baked goods, candy, coffee, cocoa, nuts, savory, and meat dishes. It has coffee, cocoa, meat, and a rubbery aroma and flavor .

Application

2-ethyl-4-methyl-1,3-thiazole is:
  • Safe and effective flavoring agent for use in animal feed, provided it is used at recommended levels and guidelines .
  • Key aroma compound in chicken soup stock.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safety and efficacy of thiazoles, thiophene and thiazoline belonging to chemical group 29 when used as flavourings for all animal species
EFSA Panel on Additives and Products or Substances used in Animal Feed (FEEDAP)
EFSA Journal, 14(6), e04441-e04441 (2016)
Aroma chemicals II: heterocycles
Zviely M
Chemistry and Technology of Flavors and Fragrances., 85-85 (2004)
Characterization of the key aroma compounds in chicken soup stock using aroma extract dilution analysis
Takakura Y, et al.
Food Science and Technology Research, 20(1), 109-113 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service