769762
Resomer® C 209, Poly(caprolactone)
ester terminated
Synonym(s):
Resomer® C 209, PCL
About This Item
Recommended Products
Quality Level
form
chunks
viscosity
0.8-1.0 dL/g
mp
56-84 °C
storage temp.
2-8°C
InChI
1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2
InChI key
PAPBSGBWRJIAAV-UHFFFAOYSA-N
Related Categories
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service