Recommended Products
isotopic purity
98 atom % D
Assay
97% (CP)
form
liquid
density
0.930 g/mL at 20 °C
mass shift
M+5
shipped in
dry ice
storage temp.
−20°C
SMILES string
[2H]C([2H])(OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)C([2H])(OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)C([2H])([2H])OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
InChI
1S/C57H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16-21,25-30,54H,4-15,22-24,31-53H2,1-3H3/b19-16-,20-17-,21-18-,28-25-,29-26-,30-27-/i52D2,53D2,54D
InChI key
HBOQXIRUPVQLKX-ADVOAEEISA-N
Packaging
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service