Skip to Content
Merck
All Photos(2)

Key Documents

704946

Sigma-Aldrich

SPhos Pd G1, Methyl t-Butyl Ether Adduct

Synonym(s):

(2-Dicyclohexylphosphino-2′,6′-dimethoxy-1,1′-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) chloride - methyl-t-butyl ether adduct, (SPhos) palladium(II) phenethylamine chloride (1:1 MTBE solvate), Chloro(2-dicyclohexylphosphino-2′,6′-dimethoxy-1,1′-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) - methyl-t-butyl ether adduct, SPhos Palladacycle, SPhos precatalyst

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H45ClNO2PPd · CH3OC4H9
Molecular Weight:
760.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

feature

generation 1

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

160-170 °C

functional group

phosphine

SMILES string

COC(C)(C)C.NCCc1ccccc1[Pd]Cl.COc2cccc(OC)c2-c3ccccc3P(C4CCCCC4)C5CCCCC5

InChI

1S/C26H35O2P.C8H10N.C5H12O.ClH.Pd/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;9-7-6-8-4-2-1-3-5-8;1-5(2,3)6-4;;/h9-11,16-21H,3-8,12-15H2,1-2H3;1-4H,6-7,9H2;1-4H3;1H;/q;;;;+1/p-1

InChI key

CLLOZWOLUBODPC-UHFFFAOYSA-M

Application

Legal Information

Usage subject to US Patents 6307087 and 6395916.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David S Surry et al.
Angewandte Chemie (International ed. in English), 47(34), 6338-6361 (2008-07-30)
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses
Ruben Martin et al.
Accounts of chemical research, 41(11), 1461-1473 (2008-07-16)
The cores of many types of polymers, ligands, natural products, and pharmaceuticals contain biaryl or substituted aromatic structures, and efficient methods of synthesizing these structures are crucial to the work of a broad spectrum of organic chemists. Recently, Pd-catalyzed carbon-carbon
David S Surry et al.
Chemical science, 2(1), 27-50 (2011-01-01)
Dialkylbiaryl phosphines are a valuable class of ligand for Pd-catalyzed amination reactions and have been applied in a range of contexts. This review attempts to aid the reader in the selection of the best choice of reaction conditions and ligand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service