Skip to Content
Merck
All Photos(1)

Key Documents

283452

Sigma-Aldrich

4-Bromo-2-thiophenecarboxaldehyde

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrOS
CAS Number:
Molecular Weight:
191.05
Beilstein:
108455
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

bp

114-115 °C/11 mmHg (lit.)

mp

44-46 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cc(Br)cs1

InChI

1S/C5H3BrOS/c6-4-1-5(2-7)8-3-4/h1-3H

InChI key

PDONIKHDXYHTLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-2-thiophenecarboxaldehyde has been used:
  • as building block for the synthesis of tetrahydroisoquinolinones
  • in the synthesis of 3-[(4-bromo-2-thienyl)methylenehydrazinocarbonyl]-1H-1,2,4-triazole

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

3-[(4-Bromo-2-thienyl) methylenehydrazinocarbonyl]-1H-1, 2, 4-triazole.
Yang J-G and Pan F-Y.
Acta Crystallographica Section E, Structure Reports Online, 60(12), o2342-o2344 (2004)
M Lebl
Bioorganic & medicinal chemistry letters, 9(9), 1305-1310 (1999-05-26)
A new technique for high throughput solid phase synthesis using the centrifuge based liquid removal from readily available standard microtiterplates is described. This technique eliminates the filtration step and is therefore applicable to simultaneous processing of an unlimited number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service