Skip to Content
Merck
All Photos(1)

Documents

12341

Sigma-Aldrich

Nonatriacontane

≥90.0% (GC)

Synonym(s):

n-Nonatriacontane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)37CH3
CAS Number:
Molecular Weight:
549.05
Beilstein:
1714035
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90.0% (GC)

mp

78-82 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C39H80/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-39H2,1-2H3

InChI key

SNXOSZZZNFRFNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thickness of single crystal of nonatriacontane is 45 ± 10 A°.

Application

Nonatriacontane was used to develop a new peak alignment algorithm named distance and spectrum correlation optimization (DISCO) method to align two-dimensional gas chromatography time-of-flight mass spectrometry data for metabolomics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

c= J'Peter Sullivan
Wunderlich B.
Reproduction (Cambridge, England) (1963)
Bing Wang et al.
Analytical chemistry, 82(12), 5069-5081 (2010-05-19)
A novel peak alignment algorithm using a distance and spectrum correlation optimization (DISCO) method has been developed for two-dimensional gas chromatography time-of-flight mass spectrometry (GCxGC/TOF-MS)-based metabolomics. This algorithm uses the output of the instrument control software, ChromaTOF, as its input

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service