Merck
HPLC Analysis of Proteins by Size Exclusion on Zenix® SEC-300 Columns, Effect of Flow Rate (15 cm column)
HPLC Analysis of Proteins by Size Exclusion on Zenix® SEC-300 Columns, Effect of Flow Rate (15 cm column) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 15 cm × 7.8 mm, 3 μm particle size, stainless steel
standard
Product No.
Description
Pricing
Thyroglobulin from bovine thyroid
powder, ≥90% (agarose gel electrophoresis)
Bovine Serum Albumin
lyophilized powder, ≥96% (agarose gel electrophoresis)
Ribonuclease A from bovine pancreas
Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
CONDITIONS
column
Zenix SEC-300, 15 cm x 7.8 mm I.D., 3 μm, 300 Å particles (Z777030)
mobile phase
150 mM sodium phosphate, pH 7
flow rate
as indicated on figure
column temp.
ambient
detector
UV, 214 nm
injection
10 μL
sample
each 1 mg/mL except uracil, 2.5 mg/mL
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies