Merck
  • Comparative proteome and transcriptome analyses of embryonic stem cells during embryoid body-based differentiation.
Comparative proteome and transcriptome analyses of embryonic stem cells during embryoid body-based differentiation.
Proteomics (2009-10-29)
Ali Fathi, Mohammad Pakzad, Adele Taei, Thore C Brink, Leila Pirhaji, Guifré Ruiz, Mohammad Sharif Tabe Bordbar, Hamid Gourabi, James Adjaye, Hossein Baharvand, Ghasem Hosseini Salekdeh
ABSTRACT
Gene expression analyses of embryonic stem cells (ESCs) will help to uncover or further define signaling pathways and molecular mechanisms involved in the maintenance of self-renewal and pluripotency. We employed a 2-DE-based proteomics approach to analyze human ESC line, Royan H5, in undifferentiated cells and different stages of spontaneous differentiation (days 3, 6, 12, and 20) by embryoid body formation. Out of 945 proteins reproducibly detected on gels, the expression of 96 spots changed during differentiation. Using MS, 87 ESC-associated proteins were identified including several proteins involved in cell proliferation, cell apoptosis, transcription, translation, mRNA processing, and protein folding. Transcriptional changes accompanying differentiation of Royan H5 were also analyzed using microarrays. We developed a comprehensive data set that shows the use of human ESC lines in vitro to mimic gastrulation and organogenesis. Our results showed that proteomics and transcriptomics data are complementary rather than duplicative. Although regulation of many genes during differentiation were observed only at transcript level, modulation of several proteins was revealed only by proteome analysis.
Sigma-Aldrich
Western Blocker Solution, for HRP detection systems
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins–Peroxidase antibody produced in mouse, clone RG-16, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse