Merck
  • Overexpression of miR-101 promotes TRAIL-induced mitochondrial apoptosis in papillary thyroid carcinoma by targeting c-met and MCL-1.
Overexpression of miR-101 promotes TRAIL-induced mitochondrial apoptosis in papillary thyroid carcinoma by targeting c-met and MCL-1.
Oncotarget (2018-01-10)
Jie Zhu, Zhenjie Li
ABSTRACT
Tumor necrosis factor-related apoptosis inducing ligand (TRAIL) induces apoptosis in malignant cells, but not in normal cells. As papillary thyroid carcinoma cells broadly expressed TRAIL receptors (death receptor 4 and death receptor 5) on their surface, TRAIL is considered as a promising drug for treatment of papillary thyroid carcinoma. However, resistance to TRAIL still be a big obstacle to achieve a satisfactory effect for cancer therapy. Here, we found that overexpression of miR-101 was able to sensitize papillary thyroid carcinoma cells to TRAIL treatment
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
MISSION® esiRNA, targeting human MET