Merck
Preventive effects of
PeerJ (2018-01-16)
Mohamed R Ben Khedher, Mohamed Hammami, Jonathan R S Arch, David C Hislop, Dominic Eze, Edward T Wargent, Małgorzata A Kępczyńska, Mohamed S Zaibi
ABSTRACT
Diet-induced obese (DIO) mice were treated for five weeks with sage methanol extract (100 and 400 mg kg After two weeks of treatment, the lower dose of sage MetOH extract decreased blood glucose and plasma insulin levels during an oral glucose tolerance test (OGTT). An insulin tolerance test (ITT), performed at day 29 confirmed that sage improved insulin sensitivity. Groups treated with low dose sage and rosiglitazone showed very similar effects on OGTT and ITT. Sage also improved HOMA-IR, triglycerides and NEFA. Treatment with the low dose increased the plasma levels of the anti-inflammatory cytokines IL-2, IL-4 and IL-10 and reduced the plasma level of the pro-inflammatory cytokines IL-12, TNF-α, and KC/GRO. The GC analysis revealed the presence of two PPARs agonist in sage MetOH extract. Sage MetOH extract at low dose exhibits similar effects to rosiglitazone. It improves insulin sensitivity, inhibits lipogenesis in adipocytes and reduces inflammation as judged by plasma cytokines. Sage presents an alternative to pharmaceuticals for the treatment of diabetes and associated inflammation.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder