Merck
  • Functional screen of MSI2 interactors identifies an essential role for SYNCRIP in myeloid leukemia stem cells.
Functional screen of MSI2 interactors identifies an essential role for SYNCRIP in myeloid leukemia stem cells.
Nature genetics (2017-04-25)
Ly P Vu, Camila Prieto, Elianna M Amin, Sagar Chhangawala, Andrei Krivtsov, M Nieves Calvo-Vidal, Timothy Chou, Arthur Chow, Gerard Minuesa, Sun Mi Park, Trevor S Barlowe, James Taggart, Patrick Tivnan, Raquel P Deering, Lisa P Chu, Jeong-Ah Kwon, Cem Meydan, Javier Perales-Paton, Arora Arshi, Mithat Gönen, Christopher Famulare, Minal Patel, Elisabeth Paietta, Martin S Tallman, Yuheng Lu, Jacob Glass, Francine E Garret-Bakelman, Ari Melnick, Ross Levine, Fatima Al-Shahrour, Marcus Järås, Nir Hacohen, Alexia Hwang, Ralph Garippa, Christopher J Lengner, Scott A Armstrong, Leandro Cerchietti, Glenn S Cowley, David Root, John Doench, Christina Leslie, Benjamin L Ebert, Michael G Kharas
ABSTRACT
The identity of the RNA-binding proteins (RBPs) that govern cancer stem cells remains poorly characterized. The MSI2 RBP is a central regulator of translation of cancer stem cell programs. Through proteomic analysis of the MSI2-interacting RBP network and functional shRNA screening, we identified 24 genes required for in vivo leukemia. Syncrip was the most differentially required gene between normal and myeloid leukemia cells. SYNCRIP depletion increased apoptosis and differentiation while delaying leukemogenesis. Gene expression profiling of SYNCRIP-depleted cells demonstrated a loss of the MLL and HOXA9 leukemia stem cell program. SYNCRIP and MSI2 interact indirectly though shared mRNA targets. SYNCRIP maintains HOXA9 translation, and MSI2 or HOXA9 overexpression rescued the effects of SYNCRIP depletion. Altogether, our data identify SYNCRIP as a new RBP that controls the myeloid leukemia stem cell program. We propose that targeting these RBP complexes might provide a novel therapeutic strategy in leukemia.
Sigma-Aldrich
Anti-hnRNP Q Antibody, clone 7A11.2, clone 7A11.2, from mouse
Sigma-Aldrich
Anti-Hox A9 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture