Merck
  • KDM3 epigenetically controls tumorigenic potentials of human colorectal cancer stem cells through Wnt/β-catenin signalling.
KDM3 epigenetically controls tumorigenic potentials of human colorectal cancer stem cells through Wnt/β-catenin signalling.
Nature communications (2017-04-26)
Jiong Li, Bo Yu, Peng Deng, Yingduan Cheng, Yongxin Yu, Kareena Kevork, Sivakumar Ramadoss, Xiangming Ding, Xinmin Li, Cun-Yu Wang
ABSTRACT
Human colorectal cancer stem cells (CSCs) are tumour initiating cells that can self-renew and are highly tumorigenic and chemoresistant. While genetic mutations associated with human colorectal cancer development are well-known, little is known about how and whether epigenetic factors specifically contribute to the functional properties of human colorectal CSCs. Here we report that the KDM3 family of histone demethylases plays an important role in tumorigenic potential and survival of human colorectal CSCs by epigenetically activating Wnt target gene transcription. The depletion of KDM3 inhibits tumorigenic growth and chemoresistance of human colorectal CSCs. Mechanistically, KDM3 not only directly erases repressive H3K9me2 marks, but also helps to recruit histone methyltransferase MLL1 to promote H3K4 methylation, thereby promoting Wnt target gene transcription. Our results suggest that KDM3 is a critical epigenetic factor in Wnt signalling that orchestrates chromatin changes and transcription in human colorectal CSCs, identifying potential therapeutic targets for effective elimination of CSCs.
Sigma-Aldrich
Anti-JMJD1C Antibody, from rabbit, purified by affinity chromatography
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture