Merck
  • A novel dual luciferase based high throughput assay to monitor autophagy in real time in yeast S. cerevisiae.
A novel dual luciferase based high throughput assay to monitor autophagy in real time in yeast S. cerevisiae.
Biochemistry and biophysics reports (2017-09-29)
Piyush Mishra, Shashank Rai, Ravi Manjithaya
ABSTRACT
Macroautophagy is a cellular response to starvation wherein superfluous and damaged cytoplasmic constituents are degraded to provide energy for survival and to maintain cellular homeostasis. Dysfunctional autophagy is attributed to disease progression in several pathological conditions and therefore, autophagy has appeared as a potential pharmacological target for such conditions. In search of potential drugs that modulate autophagy, identifying small molecule effectors of autophagy is the primary step. The conventional autophagy assays have a limitation that they cannot be scaled down to a high throughput format, therefore, novel sensitive assays are needed to discover new candidate molecules. Keeping this rationale in mind, a dual luciferase based assay was developed in the yeast S. cerevisiae that could measure both selective and general autophagy in real time. Firefly and Renilla luciferase reporter genes were cloned under POT-1 promoter. Using fatty acid medium the promoter was induced and the luciferase cargo was allowed to build up. The cells were then transferred to starvation conditions to stimulate autophagy and the degradation of luciferase markers was followed with time. The assay was more sensitive than conventional assays and could be scaled down to a 384 well format using an automated system. A good Z-factor score indicated that the assay is highly suitable for High Throughput Screening (HTS) of small molecule libraries. Screening of a small molecule library with our assay identified several known and novel modulators of autophagy.
Sigma-Aldrich
Anti-Rabbit-IgG - Atto 550 antibody produced in goat
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-Renilla Luciferase Antibody, clone 5B11.2, ascites fluid, clone 5B11.2, Chemicon®
Sigma-Aldrich
Anti-Mouse IgG−Atto 550 antibody produced in goat, 1 mg/mL protein