Merck
Microencapsulation of Gulosibacter molinativorax ON4
Journal of hazardous materials (2017-05-23)
Berta N Estevinho, A Rita Lopes, Vera Sousa, Fernando Rocha, Olga C Nunes
ABSTRACT
Molinate is a thiocarbamate herbicide used in rice crop protection. As other pesticides, molinate is a recognized environmental pollutant and bio-accumulated by some wildlife forms. Gulosibacter molinativorax ON4
Product Description
Supelco
Molinate, PESTANAL®, analytical standard
Supelco
Cycloate, PESTANAL®, analytical standard