Merck
  • Degradation pathways of 1-methylphenanthrene in bacterial Sphingobium sp. MP9-4 isolated from petroleum-contaminated soil.
Degradation pathways of 1-methylphenanthrene in bacterial Sphingobium sp. MP9-4 isolated from petroleum-contaminated soil.
Marine pollution bulletin (2016-11-21)
Jianan Zhong, Lijuan Luo, Baowei Chen, Sha Sha, Qing Qing, Nora F Y Tam, Yong Zhang, Tiangang Luan
ABSTRACT
Alkylated polycyclic aromatic hydrocarbons (PAHs) are abundant in petroleum, and alkylated phenanthrenes are considered as the primary PAHs during some oil spill events. Bacterial strain of Sphingobium sp. MP9-4, isolated from petroleum-contaminated soil, was efficient to degrade 1-methylphenanthrene (1-MP). A detailed metabolism map of 1-MP in this strain was delineated based on analysis of metabolites with gas chromatograph-mass spectrometer (GC-MS). 1-MP was initially oxidized via two different biochemical strategies, including benzene ring and methyl-group attacks. Benzene ring attack was initiated with dioxygenation of the non-methylated aromatic ring via similar degradation pathways of phenanthrene (PHE) by bacteria. For methyl-group attack, mono oxygenase system was involved and more diverse enzymes were needed than that of PHE degradation. This study enhances the understanding of the metabolic pathways of alkylated PAHs and shows the significant potential of Sphingobium sp. MP9-4 for the bioremediation of alkylated PAHs contaminated environments.
Sigma-Aldrich
1-Hydroxy-2-naphthaldehyde, AldrichCPR
Sigma-Aldrich
1-Hydroxy-2-naphthoic acid, ≥97.0%
Sigma-Aldrich
2-Hydroxy-1-naphthoic acid, 98%
Sigma-Aldrich
2-Hydroxy-1-naphthaldehyde, technical grade