Merck
Indium-mediated synthesis of heterobiaryls.
The Journal of organic chemistry (2007-04-05)
Enrique Font-Sanchis, F Javier Céspedes-Guirao, Angela Sastre-Santos, Fernando Fernandez-Lazaro
ABSTRACT
The palladium-mediated coupling reaction between triorganoindium reagents and organic electrophiles is extended to the synthesis of heteroaromatic compounds. Both electron-rich and electron-poor heterocycles can act as the organic electrophile or as the organoindium derivative.
Sigma-Aldrich
4-Iodopyrazole, 99%