Merck
  • M-sec regulates polarized secretion of inflammatory endothelial chemokines and facilitates CCL2-mediated lymphocyte transendothelial migration.
M-sec regulates polarized secretion of inflammatory endothelial chemokines and facilitates CCL2-mediated lymphocyte transendothelial migration.
Journal of leukocyte biology (2015-12-25)
Sagi Barzilai, Ronnie Blecher-Gonen, Zohar Barnett-Itzhaki, Ayelet Zauberman, Yaeli Lebel-Haziv, Ido Amit, Ronen Alon
ABSTRACT
Activation of endothelial cells by IL-1β triggers the expression of multiple inflammatory cytokines and leukocyte-attracting chemokines. The machineries involved in the secretion of these inducible proteins are poorly understood. With the use of genome-wide transcriptional analysis of inflamed human dermal microvascular endothelial cells, we identified several IL-1β-induced candidate regulators of these machineries and chose to focus our study on TNF-α-induced protein 2 (myeloid-secretory). The silencing of myeloid-secretory did not affect the ability of inflamed endothelial cells to support the adhesion and crawling of effector T lymphocytes. However, the ability of these lymphocytes to complete transendothelial migration across myeloid-secretory-silenced human dermal microvascular endothelial cells was inhibited significantly. These observed effects on lymphocyte transendothelial migration were recovered completely when exogenous promigratory chemokine CXCL12 was overlaid on the endothelial barrier. A polarized secretion assay suggested that the silencing of endothelial myeloid-secretory impairs T effector transendothelial migration by reducing the preferential secretion of endothelial-produced CCL2, a key transendothelial migration-promoting chemokine for these lymphocytes, into the basolateral endothelial compartment. Myeloid-secretory silencing also impaired the preferential secretion of other endothelial-produced inflammatory chemokines, as well as cytokines, such as IL-6 and GM-CSF, into the basolateral endothelial compartment. This is the first evidence of a novel inflammation-inducible machinery that regulates polarized secretion of endothelial CCL2 and other inflammatory chemokines and cytokines into basolateral endothelial compartments and facilitates the ability of endothelial CCL2 to promote T cell transendothelial migration.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 70,000, (FITC:Glucose = 1:250)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-MYO1C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Albumin, Human Serum, Fraction V, Low Heavy Metals