Merck
  • Glycocalyx engineering reveals a Siglec-based mechanism for NK cell immunoevasion.
Glycocalyx engineering reveals a Siglec-based mechanism for NK cell immunoevasion.
Nature chemical biology (2013-12-03)
Jason E Hudak, Stephen M Canham, Carolyn R Bertozzi
ABSTRACT
The increase of cell surface sialic acid is a characteristic shared by many tumor types. A correlation between hypersialylation and immunoprotection has been observed, but few hypotheses have provided a mechanistic understanding of this immunosuppressive phenomenon. Here, we show that increasing sialylated glycans on cancer cells inhibits human natural killer (NK) cell activation through the recruitment of sialic acid-binding immunoglobulin-like lectin 7 (Siglec-7). Key to these findings was the use of glycopolymers end-functionalized with phospholipids, which enable the introduction of synthetically defined glycans onto cancer cell surfaces. Remodeling the sialylation status of cancer cells affected the susceptibility to NK cell cytotoxicity via Siglec-7 engagement in a variety of tumor types. These results support a model in which hypersialylation offers a selective advantage to tumor cells under pressure from NK immunosurveillance by increasing Siglec ligands. We also exploited this finding to protect allogeneic and xenogeneic primary cells from NK-mediated killing, suggesting the potential of Siglecs as therapeutic targets in cell transplant therapy.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, HRP conjugate, clone 4G10®, Upstate®, from mouse
Pricing and availability is not currently available.