Merck
  • FBXW7α attenuates inflammatory signalling by downregulating C/EBPδ and its target gene Tlr4.
FBXW7α attenuates inflammatory signalling by downregulating C/EBPδ and its target gene Tlr4.
Nature communications (2013-04-12)
Kuppusamy Balamurugan, Shikha Sharan, Kimberly D Klarmann, Youhong Zhang, Vincenzo Coppola, Glenn H Summers, Thierry Roger, Deborah K Morrison, Jonathan R Keller, Esta Sterneck
ABSTRACT
Toll-like receptor 4 (Tlr4) has a pivotal role in innate immune responses, and the transcription factor CCAAT/enhancer binding protein delta (C/EBPδ, Cebpd) is a Tlr4-induced gene. Here we identify a positive feedback loop in which C/EBPδ activates Tlr4 gene expression in macrophages and tumour cells. In addition, we discovered a negative feedback loop whereby the tumour suppressor FBXW7α (FBW7, Cdc4), whose gene expression is inhibited by C/EBPδ, targets C/EBPδ for degradation when C/EBPδ is phosphorylated by GSK-3β. Consequently, FBXW7α suppresses Tlr4 expression and responses to the ligand lipopolysaccharide. FBXW7α depletion alone is sufficient to augment pro-inflammatory signalling in vivo. Moreover, as inflammatory pathways are known to modulate tumour biology, Cebpd null mammary tumours, which have reduced metastatic potential, show altered expression of inflammation-associated genes. Together, these findings reveal a role for C/EBPδ upstream of Tlr4 signalling and uncover a function for FBXW7α as an attenuator of inflammatory signalling.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Anti-mTOR/FRAP (Ab-2) Mouse mAb (22C2), liquid, clone 22C2, Calbiochem®