Merck
  • Direct and practical synthesis of 2-arylbenzoxazoles promoted by activated carbon.
Direct and practical synthesis of 2-arylbenzoxazoles promoted by activated carbon.
Organic letters (2003-09-26)
Yuka Kawashita, Natsuki Nakamichi, Hirotoshi Kawabata, Masahiko Hayashi
ABSTRACT
[reaction: see text] 2-Arylbenzoxazoles were directly synthesized from substituted 2-aminophenols and aldehydes in the presence of activated carbon (Darco KB) in xylene under an oxygen atmosphere.
Sigma-Aldrich
2-Aminophenol, 99%
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular