Merck
  • Synergistic Blockade of EGFR and HER2 by New-Generation EGFR Tyrosine Kinase Inhibitor Enhances Radiation Effect in Bladder Cancer Cells.
Synergistic Blockade of EGFR and HER2 by New-Generation EGFR Tyrosine Kinase Inhibitor Enhances Radiation Effect in Bladder Cancer Cells.
Molecular cancer therapeutics (2015-01-16)
Yu-Chieh Tsai, Pei-Yin Ho, Kai-Yuan Tzen, Tsung-Fan Tuan, Wei-Lin Liu, Ann-Lii Cheng, Yeong-Shiau Pu, Jason Chia-Hsien Cheng
ABSTRACT
Blockade of EGFR has been proved useful in enhancing the effect of radiotherapy, but the advantages of new-generation EGFR tyrosine kinase inhibitors (TKI) in radiosensitization are not well known. We used two human bladder cancer cells with wild-type EGFR to study the synergism between irradiation and afatinib (an EGFR/HER2 dual kinase inhibitor) or erlotinib (an EGFR kinase inhibitor). Here, we showed that afatinib has better radiosensitizing effect than erlotinib in increasing cancer cell killing, the percentage of apoptotic cells, and DNA damage. Afatinib is also superior to erlotinib in combining radiation to decrease tumor size, inhibit glucose metabolism, and enhance apoptotic proteins in vivo. Finally, erlotinib suppressed cell growth and induced more DNA damage in bladder cancer cells transfected with HER2 shRNA, but not in control vector-treated cells. In conclusion, concomitant blockade of radiation-activated EGFR and HER2 signaling by a new-generation EGFR TKI better inhibits the growth of bladder cancer cells both in vitro and in vivo. The absence of radiosensitization by EGFR inhibition alone and the greater radiosensitizing effect of EGFR inhibitor in HER2 knocked down cells suggest the synergism between HER2 and EGFR in determining radiosensitivity. The regained radiosensitizing activity of erlotinib implies that with proper HER2 inhibition, EGFR tyrosine kinase is still a potential target to enhance radiotherapy effect in these seemingly unresponsive bladder cancer cells.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
MISSION® esiRNA, targeting human ERBB2
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Erbb2
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Suberic acid, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Propidium iodide solution