Merck
  • Dominant-negative IKAROS enhances IL-3-stimulated signaling in wild-type but not BCR-ABL1(+) mouse BA/F3 cells.
Dominant-negative IKAROS enhances IL-3-stimulated signaling in wild-type but not BCR-ABL1(+) mouse BA/F3 cells.
Experimental hematology (2015-05-09)
Sofia von Palffy, Elizabeth Bulaeva, Sonja Babovic, Nagarajan Kannan, David J H F Knapp, Lisa Wei, Connie J Eaves, Philip A Beer
ABSTRACT
Inactivating mutations in IKZF1, the gene that encodes the transcription factor IKAROS, are recurrent in poor-prognosis human B-cell leukemias, in which these mutations co-exist with BCR-ABL1 or other genetic changes that activate similar intracellular signaling pathways. However, little is known about the mechanism(s) by which loss of IKAROS activity may co-operate with BCR-ABL1 to transform lymphoid cells. To investigate this question, we used expression of a dominant-negative isoform of IKAROS (IK6) to suppress endogenous IKAROS activity in the interleukin-3 (IL-3)-dependent mouse pro-B BA/F3 cell line and in an IL-3-independent BCR-ABL1(+) derivative. We then used intracellular phospho-flow cytometry to assess the effects of BCR-ABL1 and IK6, alone and in combination, on the signaling state of the cells before and after their stimulation with IL-3. BCR-ABL1 and IK6 each produced a constitutively activated signaling phenotype and also enhanced the signaling responses of BA/F3 cells to IL-3. These effects, however, were neither equivalent nor additive, and IK6 alone was insufficient to confer the IL-3-independent growth characteristic of BCR-ABL1(+) BA/F3 cells. In addition to its effects on lymphoid cells, IK6 also induced constitutively activated signaling in a subset of myeloid leukemia cell lines. Together, these studies indicate an ability of IK6 to enhance intracellular signaling in both lymphoid and myeloid cells, but not to synergize with BCR-ABL1 in this model system.
Sigma-Aldrich
IL-3 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Interleukin-3 from mouse, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
IL-3 from mouse, Carrier free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)