Merck
  • Effect of transpiration on plant accumulation and translocation of PPCP/EDCs.
Effect of transpiration on plant accumulation and translocation of PPCP/EDCs.
Environmental pollution (Barking, Essex : 1987) (2015-01-17)
Laurel K Dodgen, Aiko Ueda, Xiaoqin Wu, David R Parker, Jay Gan
ABSTRACT
The reuse of treated wastewater for agricultural irrigation in arid and hot climates where plant transpiration is high may affect plant accumulation of pharmaceutical and personal care products (PPCPs) and endocrine disrupting chemicals (EDCs). In this study, carrot, lettuce, and tomato plants were grown in solution containing 16 PPCP/EDCs in either a cool-humid or a warm-dry environment. Leaf bioconcentration factors (BCF) were positively correlated with transpiration for chemical groups of different ionized states (p < 0.05). However, root BCFs were correlated with transpiration only for neutral PPCP/EDCs (p < 0.05). Neutral and cationic PPCP/EDCs showed similar accumulation, while anionic PPCP/EDCs had significantly higher accumulation in roots and significantly lower accumulation in leaves (p < 0.05). Results show that plant transpiration may play a significant role in the uptake and translocation of PPCP/EDCs, which may have a pronounced effect in arid and hot climates where irrigation with treated wastewater is common.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Primidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Clofibric acid, analytical standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
Primidone, analytical standard
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%
Supelco
Diuron, PESTANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%