Merck
  • High-Throughput Analytical Techniques for Multiresidue, Multiclass Determination of 653 Pesticides and Chemical Pollutants in Tea-Part IV: Evaluation of the Ruggedness of the Method, Error Analysis, and Key Control Points of the Method.
High-Throughput Analytical Techniques for Multiresidue, Multiclass Determination of 653 Pesticides and Chemical Pollutants in Tea-Part IV: Evaluation of the Ruggedness of the Method, Error Analysis, and Key Control Points of the Method.
Journal of AOAC International (2015-04-11)
Chun-Lin Fan, Yan Li, Qiao-Ying Chang, Guo-Fang Pang, Jian Kang, Jing Cao, Yan-Bing Zhao, Nan Li, Zeng-Yin Li
ABSTRACT
A 3 month study was conducted on the ruggedness of a multiresidue method for accuracy and stability. The results indicate that in terms of Youden pair ratios of 201 pesticide aged tea samples falling approximately within 1.00-1.20 of the ratio of theoretical spraying concentrations, the differences do not exceed 5% for percentages made up by ratios of the fixed values obtained by two kinds of instruments for two teas and those made up by 18 circular determinations. However, regarding two kinds of SPE cartridges, the Cleanert TPT cartridge is higher than the ENVI-CARB+primary secondary amine (PSA) cartridge by 10%. Pertaining to RSD values of "parallel samples" and whether it is green tea or Woolong tea, the percentages of RSD≤15% values of the parallel samples all exceed 88%. Whether it is the first or circular determination for two teas and analytical results from two kinds of instruments, the percentages of RSD≤15% values have a difference of less than 6%, while the TPT cartridge is better than ENVI-CARB+PSA by above 6% for the two cartridges. Concerning RSDs of Youden pair ratios, RSD≤15% values have a proportion exceeding 85% for both green tea and Woolong tea, and the percentage is greater than 87% whether it is for two kinds of SPE cartridges or two kinds of instruments. In terms of Youden pair ratios and the classified statistical analysis of the ruggedness data of parallel samples, the proportion of RSD≤15% values of Youden pair ratios is 8% higher for the TPT cartridge than the ENVI-CARB+PSA cartridge; the proportion of RSD≤15% values of parallel samples is 6.2% higher for the TPT cartridge than the ENVI-CARB+PSA cartridge. Data show no marked differences for two teas and two kinds of instruments. A comparison of the aforementioned aspects finds that good ruggedness was obtained with both SPE cleanup methods, and the results from the TPT cartridge are better than those from the ENVI-CARB+PSA cartridge.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Visiprep 5-port Flask Vacuum Manifold, DL (Disposable Liner)