Merck
  • Inhibition of the thioesterase activity of human fatty acid synthase by 1,4- and 9,10-diones.
Inhibition of the thioesterase activity of human fatty acid synthase by 1,4- and 9,10-diones.
Chemical & pharmaceutical bulletin (2014-09-02)
Herman Odens, Todd Lowther, Steven Kridel, Laura Watts, Lauren Filipponi, Jeffrey Schmitt
ABSTRACT
Fatty acid synthase (FASN) is the enzyme that synthesizes fatty acids de novo in human cells. Although FASN is generally expressed at low levels in most normal tissues, its expression is highly upregulated in many cancers. Consistent with this notion, inhibition of FASN activity has demonstrated potential to halt proliferation and induce cell death in vitro and to block tumor growth in vivo. Consequently, FASN is widely recognized as a valuable therapeutic target. In this report, we describe a variety of 1,4-quinones and 9,10-anthraquinones, including several natural compounds and some newly synthesized compounds, that potently inhibit the thioesterase (TE) domain of FASN. Inhibition of recombinant TE activity, inhibition of cellular FASN, and cytotoxicity in human prostate cancer cell lines and normal fibroblasts, is shown for the most potent inhibitors. Collectively, the data illustrate the novel inhibitory capacity of the 1,4-quinone and 9,10-anthraquinone pharmacophores against FASN.
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
Benzyltrimethylammonium bromide, 97%
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)