Merck
Tasimelteon: a selective and unique receptor binding profile.
Neuropharmacology (2014-12-24)
Christian Lavedan, Mark Forsberg, Anthony J Gentile
ABSTRACT
Hetlioz(®) (tasimelteon) is the first approved treatment in the United States for Non-24-Hour Sleep-Wake Disorder (Non-24). We present here data on the in vitro binding affinity of tasimelteon for both human melatonin receptors MT1 and MT2, as well as the extended screen of other receptors and enzymes. Results indicate that tasimelteon is a potent Dual Melatonin Receptor Agonist (DMRA) with 2.1-4.4 times greater affinity for the MT2 receptor believed to mediate circadian rhythm phase-shifting (Ki = 0.0692 nM and Ki = 0.17 nM in NIH-3T3 and CHO-K1 cells, respectively), than for the MT1 receptor (Ki = 0.304 nM and Ki = 0.35 nM, respectively). Tasimelteon was also shown to have no appreciable affinity for more than 160 other pharmacologically relevant receptors and several enzymes.
Product Description
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Tasimelteon, ≥98% (HPLC)