Merck
  • Characterization of virus-like particles in GARDASIL® by cryo transmission electron microscopy.
Characterization of virus-like particles in GARDASIL® by cryo transmission electron microscopy.
Human vaccines & immunotherapeutics (2013-12-05)
Qinjian Zhao, Clinton S Potter, Bridget Carragher, Gabriel Lander, Jaime Sworen, Victoria Towne, Dicky Abraham, Paul Duncan, Michael W Washabaugh, Robert D Sitrin
ABSTRACT
Cryo-transmission electron microscopy (cryoTEM) is a powerful characterization method for assessing the structural properties of biopharmaceutical nanoparticles, including Virus Like Particle-based vaccines. We demonstrate the method using the Human Papilloma Virus (HPV) VLPs in GARDASIL®. CryoTEM, coupled to automated data collection and analysis, was used to acquire images of the particles in their hydrated state, determine their morphological characteristics, and confirm the integrity of the particles when absorbed to aluminum adjuvant. In addition, we determined the three-dimensional structure of the VLPs, both alone and when interacting with neutralizing antibodies. Two modes of binding of two different neutralizing antibodies were apparent; for HPV type 11 saturated with H11.B2, 72 potential Fab binding sites were observed at the center of each capsomer, whereas for HPV 16 interacting with H16.V5, it appears that 60 pentamers (each neighboring 6 other pentamers) bind five Fabs per pentamer, for the total of 300 potential Fab binding sites per VLP.
Sigma-Aldrich
L-Cysteine hydrochloride hydrate, 99%
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, Wacker Chemie AG, 98.5-101.0%
Supelco
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, JIS special grade, ≥99.0%
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard