Merck
  • Quantum dot solar cells. harvesting light energy with CdSe nanocrystals molecularly linked to mesoscopic TiO2 films.
Quantum dot solar cells. harvesting light energy with CdSe nanocrystals molecularly linked to mesoscopic TiO2 films.
Journal of the American Chemical Society (2006-02-16)
István Robel, Vaidyanathan Subramanian, Masaru Kuno, Prashant V Kamat
ABSTRACT
By using bifunctional surface modifiers (SH-R-COOH), CdSe quantum dots (QDs) have been assembled onto mesoscopic TiO(2) films. Upon visible light excitation, CdSe QDs inject electrons into TiO(2) nanocrystallites. Femtosecond transient absorption as well as emission quenching experiments confirm the injection from the excited state of CdSe QDs into TiO(2) nanoparticles. Electron transfer from the thermally relaxed s-state occurs over a wide range of rate constant values between 7.3 x 10(9) and 1.95 x 10(11) s(-1). The injected charge carriers in a CdSe-modified TiO(2) film can be collected at a conducting electrode to generate a photocurrent. The TiO(2)-CdSe composite, when employed as a photoanode in a photoelectrochemical cell, exhibits a photon-to-charge carrier generation efficiency of 12%. Significant loss of electrons occurs due to scattering as well as charge recombination at TiO(2)/CdSe interfaces and internal TiO(2) grain boundaries.
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Pricing and availability is not currently available.