Merck
The pathophysiology of elevated vitamin B12 in clinical practice.
QJM : monthly journal of the Association of Physicians (2013-03-01)
E Andrès, K Serraj, J Zhu, A J M Vermorken
ABSTRACT
Hypercobalaminemia (high serum vitamin B12 levels) is a frequent and underestimated anomaly. Clinically, it can be paradoxically accompanied by signs of deficiency, reflecting a functional deficiency linked to qualitative abnormalities, which are related to defects in tissue uptake and action of vitamin B12. The aetiological profile of high serum cobalamin predominantly encompasses severe disease entities for which early diagnosis is critical for prognosis. These entities are essentially comprised of solid neoplasms, haematological malignancies and liver and kidney diseases. This review reflects the potential importance of the vitamin B12 assay as an early diagnostic marker of these diseases. A codified approach is needed to determine the potential indications of a search for high serum cobalamin and the practical clinical strategy to adopt upon discovery of elevated cobalamin levels. While low serum cobalamin levels do not necessarily imply deficiency, an abnormally high serum cobalamin level forms a warning sign requiring exclusion of a number of serious underlying pathologies. Functional cobalamin deficiency can thus occur at any serum level.
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material