Merck
  • A rapid approach for the detection, quantification, and discovery of novel sulfenic acid or S-nitrosothiol modified proteins using a biotin-switch method.
A rapid approach for the detection, quantification, and discovery of novel sulfenic acid or S-nitrosothiol modified proteins using a biotin-switch method.
Methods in enzymology (2010-06-02)
Joseph R Burgoyne, Philip Eaton
ABSTRACT
The recent development of robust methods for the detection of proteins susceptible to S-nitrosylation (RSNO) and sulfenation (RSOH) has provided greater insight into the role of these oxidative modifications in cell signaling. These techniques, which have been termed "biotin-switch" methods, essentially use selective chemical reduction to swap an oxidative modification for a stable easily detectable biotin-tag. This allows for the rapid purification and subsequent detection of modified proteins using mass spectrometry. This chapter provides an overview of these biotin-switch methods, and explores its impact on the field of redox biology, including recent advances as well as limitations associated with this technique.
Product Description
SAFC
BIOTIN
Sigma-Aldrich
Biotin, SAJ special grade, ≥98.0%
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Biotin, United States Pharmacopeia (USP) Reference Standard