Merck
The role of glimepiride in the effective management of Type 2 diabetes.
Journal of diabetes and its complications (2004-11-09)
Stephen N Davis
ABSTRACT
Type 2 diabetes mellitus, a disorder of impaired insulin secretion and insulin resistance, has reached epidemic proportions. The effective management of Type 2 diabetes is of vital concern to clinicians. The identification of high-risk individuals and lifestyle management can help control diabetes; however, most patients require pharmacologic intervention. The goals of pharmacologic therapy are to achieve adequate glycemic control while avoiding hypoglycemia and weight gain and to minimize the risk of future micro- and macrovascular complications. There are a number of available glucose-lowering agents from which to choose. This review focuses on the sulfonylureas, the first oral agents introduced for the management of Type 2 diabetes, which are effective, well-tolerated, and well-established drugs, Second-generation sulfonylureas are now widely used in the management of Type 2 diabetes. The most recent addition, glimepiride, can be used in combination with metformin, the thiazolidinediones, alpha-glucosidase inhibitors, and insulin. The unique properties of glimepiride may provide advantages over other currently available insulin secretagogues.
Product Description
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid
USP
Glimepiride, United States Pharmacopeia (USP) Reference Standard
Glimepiride, European Pharmacopoeia (EP) Reference Standard