Merck
  • Review of the background and application of triolein-containing semipermeable membrane devices in aquatic environmental study.
Review of the background and application of triolein-containing semipermeable membrane devices in aquatic environmental study.
Aquatic toxicology (Amsterdam, Netherlands) (2002-09-03)
Yibing Lu, Zijian Wang, James Huckins
ABSTRACT
This paper briefly reviews research on passive in situ samplers for aquatic environments but focuses on the development and application of the triolein-containing semipermeable membrane device in aquatic environmental monitoring. Special attention is paid to the calibration of the devices, quality control issues, and its potential uses in environmental assessments of aquatic contaminants. Also, the suitability of the technique for incorporation with selected bioassays is examined.
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Glyceryl trioleate, ≥97.0% (TLC)
Sigma-Aldrich
Glyceryl trioleate, ~65%
Supelco
Glyceryl trioleate, analytical standard
Triolein, European Pharmacopoeia (EP) Reference Standard