Merck
  • Development and validation of ethylenethiourea determination in foods using methanol-based extraction, solid-phase extraction cleanup and LC-MS/MS.
Development and validation of ethylenethiourea determination in foods using methanol-based extraction, solid-phase extraction cleanup and LC-MS/MS.
Food chemistry (2013-04-13)
Kevin Tran, Lucie P Mactal, Michele R Cromer, Ralph H Vocque, Robert E Smith
ABSTRACT
To response to the need for a rapid, cost-effective, eco-friendly and efficient extraction process, a sensitive method for the determination of ethylenethiourea (ETU) in food matrices by high-performance liquid chromatography-mass spectrometry (HPLC-LC/MS) was developed and validated. ETU was extracted from food commodities with methanol, cleaned up by alumina-SPE column, and then determined by HPLC-MS/MS. The MS detection was operated in positive ionization mode. For confirmation of target compound, two precursor ion>product ion transitions were selected by multi-reaction monitoring mode (MRM). The method showed good linearity with correlation coefficient higher than 0.9950. Recoveries at three spiked levels (10, 50, 100 ng/g) in random selected food matrices were in range of 71-121% with RSDs not larger than 25%. The limit of quantitation for the analyte was estimated at 5 ng/g.
Product Description
Supelco
2-Imidazolidinethione, PESTANAL®, analytical standard
Sigma-Aldrich
2-Imidazolidinethione, 98%
Sigma-Aldrich
2-Imidazolidinethione, purum, ≥98.0% (HPLC)