- Radiation damage due to knock-on processes on carbon foils cooled to liquid helium temperature.
Radiation damage due to knock-on processes on carbon foils cooled to liquid helium temperature.
Ultramicroscopy (1978-01-01)
I Dietrich, F Fox, H G Heide, E Knapek, R Weyl
PMID695132
ABSTRACT
Radiation damage on a holey carbon foil was investigated in an electron microscope with a superconducting lens system, where the temperature of the specimen and its environment initially was 4 K. Due to an electron dose of 2 X 10(4) As/cm2 the diameter of a hole increased 5 nm. Rough calculations show that this increase can be ascribed to knock-on processes. Estimates of the rise in specimen temperature during the irradiation are given.
Product Description
Carbon, foil, 100x100mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 250x250mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 50x50mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 250x250mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 150x150mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 150x150mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 25x25mm, thickness 0.1mm, pyrolytic graphite, 99.99%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon, foil, 300x300mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon, foil, not light tested, 100x100mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 150x150mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 25x25mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 150x150mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 12mm disks, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 100x100mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%