Merck
  • Gender-related and age-related urinalysis of healthy subjects by NMR-based metabonomics.
Gender-related and age-related urinalysis of healthy subjects by NMR-based metabonomics.
NMR in biomedicine (2007-05-03)
Nikolaos G Psihogios, Irene F Gazi, Moses S Elisaf, Konstantin I Seferiadis, Eleni T Bairaktari
ABSTRACT
NMR-based metabonomic analysis is a well-established approach to characterizing healthy and diseased states. The aim of this study was to investigate inter-individual variability in the metabolic urinary profile of a healthy Greek population, not subjected to strict dietary limitations, by NMR-based metabonomics. The overall metabonomic urinalysis showed a homogeneous distribution among the population. The metabolic profile was examined in relation to gender and age, and reference intervals of major metabolites were determined. Multivariate data analysis led to the construction of two robust models that were able to predict the class membership of the subjects studied according to their gender and age. The most influential low molecular weight metabolites responsible for the differences in gender groups were citrate, creatinine, trimethylamine N-oxide, glycine, creatine and taurine, and for the differences in age groups they were citrate, creatinine, trimethylamine N-oxide and an unidentified metabolite (delta 3.78).
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
L-Alanine, ≥99%
SAFC
Glycine
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Succinic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Dimethylamine solution, CP, 50% in H2O
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C