Merck
Continuing methadone for pain in palliative care.
Pain research & management (2013-05-11)
Philippa Hawley, Ryan Liebscher, Jessica Wilford
ABSTRACT
Methadone is one of the most important medications used for the treatment of refractory pain in the palliative care setting, and is usually initially prescribed by one of a limited number of physicians who have acquired authorization for its use. A lack of authorized physicians able to take over prescribing when the patient is stable is a barrier to accessing methadone for analgesia. To determine the barriers to family physicians becoming authorized to prescribe methadone for pain in palliative care. A survey exploring the perceived barriers to continuing methadone for pain in palliative care following initial prescription by a specialist was mailed to a randomly selected group of 870 family physicians in British Columbia. The response rate was 30.9%. Of the 204 responding physicians, 76.1% described themselves as positioned to provide ongoing palliative care to their patients. Within this group, 38 (18.6%) were already authorized to prescribe methadone for pain. The remaining 166 (81.4%) had significant knowledge deficits regarding methadone use in palliative care, but were largely aware of their deficits, and more than one-half were willing to learn more and to obtain an authorization if requested. Responding family physicians had mostly received little education regarding methadone for pain, but were aware of their need for education and were willing to learn. Physicians who had already become authorized were generally satisfied with the process of authorization, and believed the process of education through authorization was appropriate and not onerous.
Sigma-Aldrich
(±)-Methadone hydrochloride, powder, ≥98%
Supelco
(±)-Methadone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®