Merck
Inorganic pyrophosphatases: one substrate, three mechanisms.
FEBS letters (2013-05-21)
Tommi Kajander, Juho Kellosalo, Adrian Goldman
ABSTRACT
Soluble inorganic pyrophosphatases (PPases) catalyse an essential reaction, the hydrolysis of pyrophosphate to inorganic phosphate. In addition, an evolutionarily ancient family of membrane-integral pyrophosphatases couple this hydrolysis to Na(+) and/or H(+) pumping, and so recycle some of the free energy from the pyrophosphate. The structures of the H(+)-pumping mung bean PPase and the Na(+)-pumping Thermotoga maritima PPase solved last year revealed an entirely novel membrane protein containing 16 transmembrane helices. The hydrolytic centre, well above the membrane, is linked by a charged "coupling funnel" to the ionic gate about 20Å away. By comparing the active sites, fluoride inhibition data and the various models for ion transport, we conclude that membrane-integral PPases probably use binding of pyrophosphate to drive pumping.
Sigma-Aldrich
Pyrophosphoric acid, technical grade
Sigma-Aldrich
Pyrophosphoric acid, SAJ first grade, ≥90.0%