Merck
  • Intranasal administration of antibody-bound respiratory syncytial virus particles efficiently primes virus-specific immune responses in mice.
Intranasal administration of antibody-bound respiratory syncytial virus particles efficiently primes virus-specific immune responses in mice.
Journal of virology (2013-05-03)
Debby Kruijsen, Helga K Einarsdottir, Marcel A Schijf, Frank E Coenjaerts, Ellen C van der Schoot, Gestur Vidarsson, Grada M van Bleek
ABSTRACT
Infants are protected from a severe respiratory syncytial virus (RSV) infection in the first months of life by maternal antibodies or by prophylactically administered neutralizing antibodies. Efforts are under way to produce RSV-specific antibodies with increased neutralizing capacity compared to the currently licensed palivizumab. While clearly beneficial during primary infections, preexisting antibodies might affect the onset of adaptive immune responses and the ability to resist subsequent RSV infections. Therefore, we addressed the question of how virus neutralizing antibodies influence the priming of subsequent adaptive immune responses. To test a possible role of the neonatal Fc receptor (FcRn) in this process, we compared the responses in C57BL/6 wild-type (WT) and FcRn(-/-) mice. We observed substantial virus-specific T-cell priming and B-cell responses in mice primed with RSV IgG immune complexes resulting in predominantly Th1-type CD4(+) T-cell and IgG2c antibody responses upon live-virus challenge. RSV-specific CD8(+) T cells were primed as well. Activation of these adaptive immune responses was independent of FcRn. Thus, neutralizing antibodies that localize to the airways and prevent infection-related routes of antigen processing can still facilitate antigen presentation of neutralized virus particles and initiate adaptive immune responses against RSV.
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Picric acid solution
Sigma-Aldrich
Picric acid, moistened with water, ≥98%