Merck
Lead monoxide α-PbO: electronic properties and point defect formation.
Journal of physics. Condensed matter : an Institute of Physics journal (2013-01-24)
J Berashevich, O Semeniuk, O Rubel, J A Rowlands, A Reznik
ABSTRACT
The electronic properties of polycrystalline lead oxide consisting of a network of single-crystalline α-PbO platelets and the formation of native point defects in the α-PbO crystal lattice are studied using first-principles calculations. The results suggest that the polycrystalline nature of α-PbO causes the formation of lattice defects (i.e., oxygen and lead vacancies) in such a high concentration that defect related conductivity becomes the dominant mechanism of charge transport. The neutral O vacancy forms a defect state at 1.03 eV above the valence band which can act as a deep trap for electrons, while the Pb vacancy forms a shallow trap for holes located just 0.1 eV above the valence band. The ionization of O vacancies can account for the experimentally found dark current decay in ITO/PbO/Au structures.
Sigma-Aldrich
Lead(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) oxide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Lead(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Lead(II) oxide, puriss. p.a., ≥99.0% (KT), yellow
Sigma-Aldrich
Lead(II) oxide, powder, <10 μm, ReagentPlus®, ≥99.9% trace metals basis