Merck
  • Adduct formation identification between phenyl glycidyl ether and 2'-deoxyadenosine and thymidine by chromatography, mass spectrometry and nuclear magnetic resonance spectroscopy.
Adduct formation identification between phenyl glycidyl ether and 2'-deoxyadenosine and thymidine by chromatography, mass spectrometry and nuclear magnetic resonance spectroscopy.
Journal of chromatography (1990-03-30)
E Van den Eeckhout, A De Bruyn, H Pepermans, E L Esmans, I Vryens, J Claereboudt, M Claeys, J E Sinsheimer
ABSTRACT
Thymidine and 2'-deoxyadenosine were reacted with phenyl glycidyl ether in order to study the formation of the corresponding 2'-deoxynucleoside adducts. Separation methods were elaborated using either reversed-phase high-performance liquid chromatography with photodiode-array detection, or centrifugal circular thin-layer chromatography. The adducts were isolated on a preparative scale and were fully characterized by UV spectroscopy, desorption chemical ionization and fast atom bombardment mass spectrometry and 270- and 360-MHz 1H NMR spectrometry. For thymidine the main adduct was characterized as N-3-(2-hydroxy-3-phenoxypropyl)thymidine. With 2'-deoxyadenosine, predominantly N-1-(2-hydroxy-3-phenoxypropyl)-2'-deoxyadenosine was formed. With longer reaction times, the formation of a minor amount of dialkylated 2'-deoxyadenosine was observed. These nucleoside adducts will be used as marker compounds for studies of DNA adduct formation.
Sigma-Aldrich
1,2-Epoxy-3-phenoxypropane, technical grade
Sigma-Aldrich
1,2-Epoxy-3-phenoxypropane, 99%