Merck
  • Kinetic, isotherm and thermodynamic studies of the adsorption of crystal violet by activated carbon from peanut shells.
Kinetic, isotherm and thermodynamic studies of the adsorption of crystal violet by activated carbon from peanut shells.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-01-12)
J X Zhang, L L Ou
ABSTRACT
The adsorption of crystal violet dye from aqueous solutions onto an activated carbon prepared from peanut shells was analyzed in this study. The effects of particle size, initial concentration, time and temperature on crystal violet removal were studied in batch experiments. Experimental results showed that the adsorption equilibrium was achieved within 100 min for all studied concentrations. Analysis of adsorption results showed that the adsorption isotherms could be well fitted to the Langmuir model. Kinetic parameters, rate constants, equilibrium adsorption capacities and related correlation coefficients for pseudo first-order and second-order kinetic models were calculated and discussed. The results revealed that the adsorption kinetics was in good agreement with the pseudo second-order equation. Thermodynamic parameters such as the change of Gibbs free energy (ΔG°), change of enthalpy (ΔH°) and change of entropy (ΔS°) have also been determined and it has been found that the adsorption process should be spontaneous, endothermic and physisorption in nature.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Gram-Hucker′s stain solution (I)
Sigma-Aldrich
Gentian Violet solution
Sigma-Aldrich
Crystal Violet, JIS special grade
Sigma-Aldrich
Crystal Violet, SAJ first grade
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet Solution