Merck
  • Control of phosphoryl migratory transesterifications allows regioselecive access to sugar phosphates.
Control of phosphoryl migratory transesterifications allows regioselecive access to sugar phosphates.
Organic letters (2013-01-05)
Mitul K Patel, Benjamin G Davis
ABSTRACT
Phosphate esters in polyhydroxylated systems are normally blighted by uncontrolled migration under a variety of reaction conditions. Cesium fluoride is demonstrated as a reagent to control migration of primary phosphates during transesterifications. This allows easy exchange of phosphoryl protecting groups enabling enhanced synthetic strategic flexibility and regioselective phosphate installation. Mechanistic analysis suggests that a fluoride-induced extended solvent sphere modulates steric bulk at phosphorus to favor the primary position.
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Sigma-Aldrich
Cesium Fluoride ChemBeads, NSC 84270