Merck
  • Cryopreservation of rabbit semen: comparing the effects of different cryoprotectants, cryoprotectant-free vitrification, and the use of albumin plus osmoprotectants on sperm survival and fertility after standard vapor freezing and vitrification.
Cryopreservation of rabbit semen: comparing the effects of different cryoprotectants, cryoprotectant-free vitrification, and the use of albumin plus osmoprotectants on sperm survival and fertility after standard vapor freezing and vitrification.
Theriogenology (2012-12-12)
Maria Pina Rosato, Nicolaia Iaffaldano
ABSTRACT
This study was designed to improve current freezing protocols for rabbit sperm by examining: (1) the toxicity of different permeable cryoprotectants (CPAs) used for standard vapor freezing (conventional freezing); (2) the feasibility of ultrarapid nonequilibrium freezing (vitrification) of sperm in the absence of permeating CPAs; and (3), the addition of bovine serum albumin (BSA), alone or with sucrose or trehalose as osmoprotectants. First, we evaluated the effects on sperm motility of the incubation time (5 to 60 minutes) with different final concentrations (5% to 20%) of glycerol, N-N-dimethylacetamide, dimethylsulfoxide (DMSO), ethylene glycol, propylene glycol, and methanol. N-N-dimethylacetamide (5%) and DMSO (5% and 10%) showed the least toxic effects; the use of 10% DMSO producing the best postthaw sperm motility and membrane integrity results (P < 0.05) after conventional freezing. For vitrification, semen was diluted in the absence of permeable CPAs and frozen by dropping semen directly in liquid nitrogen. However, this led to the low or null cryosurvival of sperm postvitrification (0.16 ± 0.4%, 1.8 ± 1.6%, and 94.5 ± 1.4% of motile, membrane-, and DNA-intact sperm cells, respectively). To assess the effects of albumin and osmoprotectants on sperm cryosurvival, sperm was conventionally frozen with 10% DMSO or vitrified in the absence of permeable CPAs without or with 0.5% BSA alone or combined with sucrose or trehalose (range, 0-0.25 M). In the conventional freezing procedure, the addition of BSA alone failed to improve sperm cryosurvival, however, in the presence of BSA plus either sucrose or trehalose, the postthaw motility (using 0.1 M sucrose or trehalose) and DNA integrity (using all additive concentrations) of sperm were significantly better (P < 0.05) than control. Higher numbers of motile and membrane-intact cells were observed when semen was vitrified with BSA alone or with BSA and sucrose (0.1 and 0.25 M) or BSA and trehalose (0.25 M) and a best recovery of DNA-intact sperm was recorded for BSA plus sucrose compared with semen vitrified without osmoprotectants (P < 0.05). Finally, the cryodiluent combinations BSA/sucrose and BSA/trehalose were compared in an insemination trial. Rabbit does were inseminated with fresh semen (N = 56), semen conventionally cryopreserved in the BSA-based cryodiluents containing 0.1 M sucrose or trehalose (N = 56 per group), or semen vitrified in the presence of 0.25 M sucrose or trehalose (N = 8 per group). Fertility rates and live born kids were similar for semen cryopreserved with BSA/sucrose (77% and 7.6) compared with fresh semen (84% and 8.1) and significantly higher than the figures recorded for the conventionally frozen semen in the BSA/trehalose group (52% and 6.1; P ≤ 0.05). In contrast, only one doe inseminated with semen vitrified in the presence of BSA/sucrose became pregnant, though no kids were delivered. The conclusions to be drawn from our study are: (1) incubation times and concentration toxicities established for the main permeable CPAs used for conventional freezing of rabbit sperm indicated that DMSO 10% was the least damaging; (2) CPA-free vitrification of rabbit semen led to a low or null sperm cryosurvival; and (3) enriching the freezing medium with BSA plus adequate amounts of sucrose or trehalose can improve the cryosurvival of rabbit sperm after conventional freezing or vitrification. In our working conditions, BSA/sucrose was more effective than BSA/trehalose at preserving the in vivo fertilization capacity of rabbit sperm cryopreserved using the standard procedure.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
(S)-(+)-1,2-Propanediol, 96%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%